Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Analyser
Cellulär analys
EZAssay™ TBARS Estimation Kit for Lipid Peroxidation
EZAssay™ TBARS Estimation Kit for Lipid Peroxidation
Katalog # HIMECCK023-100
Leverantör:
HIMEDIA
New Product
CAS-nummer:
Expandera bilden
EZAssay™ TBARS Estimation Kit for Lipid Peroxidation
Katalog # HIMECCK023-100
Leverantör:
HIMEDIA
New Product
CAS-nummer:
Specifikationer
Förp.:
1 KIT
Beskrivning:
EZAssay™ TBARS estimation kit for lipid peroxidation