Magicard™ Prima 8 Transparent Laminate Film with Chip Recess
Katalog # BRDY321994
Leverantör: Brady
New Product
Specifikationer
- Beskrivning:Prima 8 transparent laminate film with chip recess
- Förp.:1
Specifikationer
Om denna produkt
The Prima 8 transparent laminate film with a chip recess offers extra protection against scratches on your card and can be applied using the Prima 8 laminator.
- Print technology: Thermal transfert
- Printer compatibility: Prima 8 laminator
Applications: General identification, security identification, healthcare and hospital labels and site identification.