TouchNTuff® 93-300 Disposable Nitrile Cleanroom Gloves, Non Sterile
Supplier: Ansell
New Product
Certificates
About this item
TouchNTuff® 93-300 nitrile cleanroom gloves are comfortable and robust, ensuring strong chemical splash resistance and resistance against punctures and tears.
- Robust nitrile formulation: Puncture, tear and chemical splash resistance
- Silicone-free design: Improved product protection
- Class 100 (ISO 5) cleanroom compatibility: Assured cleanliness
- Suitable for double gloving
- Powder-free, chlorinated
- Tested for use with chemotherapy drugs
These nitrile gloves are entirely silicone-free, preventing potential contaminant transfer and safeguarding product integrity.
They are also designed to be compatible with Class 100 (ISO 5) cleanroom environments, delivering hand protection that is reliable and fit for purpose.
Recommended for cleanroom cleaning and preparation, blending and compounding of solids and liquids, spill or leakage cleanup, transferring liquids and solids, loading centrifuges and chromatography columns, part assembly, and weighing and dispensing of solid and liquid raw materials.
Certifications: EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2); Cat. III
Food contact approved (FDA)
Food contact approved (FDA)
Packaging: 50 gloves per vacuum sealed polybag; 2 polybags per master polybag; 10 master poly bags per poly lined case.
Specifications
- Catalog No:
- ANSE93-300/5.5-6
- ANSE93-300/9.5-10
- ANSE93-300/8.5-9
- ANSE93-300/7.5-8
- ANSE93-300/6.5-7
- Modell:
- 93-300
- 93-300
- 93-300
- 93-300
- 93-300
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Renrumsklass:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Material:
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Finger, tjocklek:
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- Handflata, tjocklek:
- 0,13 mm
- 0,13 mm
- 0,13 mm
- 0,13 mm
- 0,13 mm
- FDA-reglerad:
- +
- +
- +
- +
- +
- EN standard:
- EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2)
- EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2)
- EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2)
- EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2)
- EN ISO 374-1:2016 (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (Part 2)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Antistatisk:
- +
- +
- +
- +
- +
- ASTM nr.:
- D6319
- D6319
- D6319
- D6319
- D6319