Magicard™ Monochrome Black Film + Overcoat for Magicard™ 600
BRDY321970
: Brady
New Product
- Färg:Genomskinlig/svart
- Beskrivning:Black monochrome film with overcoat for Magicard™ 600 printer
- Längd:12 m
- Förp.:1
- Bredd:60 mm
- För:Magicard™ 600
Magicard™ 600 black monochrome ribbon with overcoat.
- Create extra durable cards
- Includes a cleaning roller for dust free printing
Black mono colour dye film with overcoat protection.
: Includes 1 dye film and 1 cleaning roller.