Tegaderm™ Transparent Film Roll
Katalog # MMMA7100221528
Leverantör: 3M
New Product
Specifikationer
- Beskrivning:Transparent film roll
- L×B:10 m × 150 mm
- Förp.:1 Roll
Specifikationer
Om denna produkt
Non-sterile version of Tegaderm™ Transparent film roll provides all the benefits of surgical tape while providing a waterproof, bacterial and viral barrier.
- The conforming film moves with the skin for greater patient comfort
- Breathable foil allows oxygen and steam to pass through
- It can be cut and adjusted to the required length and shape
- Easy to apply with gloves on
- Waterproof
- Industrial category: Safety
In vitro testing shows that the transparent film provides a viral barrier from viruses 27 nm in diameter or larger while the dressing remains intact without leakage. The frame makes application precise and easy, minimising the risk of the dressing sticking and the possibility of sticking to gloves.