Neo-Sensitabs™
: ROSCO
Neo-Sensitabs are tablets manufactured by a process using crystalline antimicrobials. Extensive range - over 125 different antimicrobial agents and combination tablets are available. Detection of mechanisms of resistance including MRSA, ESBL, AmpC Inducers and Inducible Clindamycin resistance. Logical 5-figure alpha-numeric code, easy and safe identification of the individual tablet, optically readable by automatic reader systems. Neo-Sensitabs are standardized to CLSI recommendations and adjusted to other national guidelines.
- Long shelf life - up to five years
- Large easy-to-read printed codes
- Uniform and reliable results at all times, no loss of activity due to degradation of the antibiotics
- Store and use at room temperature without degradation of the antimicrobial agent