Mini Drum
Mini drums, 6 L, Rostfritt stål 304, 175 mm
Katalog # SAMPA434-6
Leverantör: Sampling Systems
Gå till:
Produktdetaljer och dokument
Mini drums make for ideal storage and production containers.
- Made from high-quality 304 stainless steel
- Ideal for both powders and liquids
- Supplied with a drop-on lid
- Toggle clamps and O-ring seals are optional extras
- Available from stock
- Fast delivery
This range of mini drums are ideal for the storage of both powders and liquids. They come complete with a drop-on lid. Optional extras include toggle clamps and O-ring seals, enabling the lid to be firmly secured.
Specifikationer
- Ø utv.:175 mm
- Kapacitet:6 L
- Beskrivning:Mini drums
- Material:Rostfritt stål 304