- Förp.:1 KIT
- Beskrivning:ImmunoCult™ Human B cell expansion kit
Serum-free culture kit for expansion of human B cells.
Reliably expand human B cells that are ready for use in downstream assays and applications - without the use of serum, specialized cultureware, or problematic feeder cells. ImmunoCult™ Human B Cell Expansion Kit comprises ImmunoCult™-ACF Human B Cell Expansion Supplement and ImmunoCult™-XF B Cell Base Medium, which together ensure consistent activation and expansion of human B cells and their maturation to plasma cells. B cells can be harvested and used directly in downstream applications after just one week or less in culture.
For convenience, all kit components are also available for individual purchase. This kit is compatible with many other upstream and downstream products. For example, use EasySep™ cell separation kits to isolate a range of B cell subsets - such as pan-B cells, memory B cells, and naïve B cells - that can be isolated and then immediately expanded using ImmunoCult™ Human B Cell Expansion Kit.