ContactUsLinkComponent
STEMdiff™ Pancreatic Progenitor Kit
STEMdiff™ Pancreatic Progenitor Kit
Catalog # STMC05120
CAS Number:  
undefined
STEMdiff™ Pancreatic Progenitor Kit
Catalog # STMC05120
CAS Number:  
Specifications
  • Förp.:
    1 KIT
  • Beskrivning:
    STEMdiff™ Pancreatic progenitor kit
Specifications
About this item
Serum-free medium for the differentiation of human ES and iPS cells to pancreatic progenitor cells.
STEMdiff™ Pancreatic Progenitor Kit is a serum-free medium that supports efficient and reproducible generation of pancreatic progenitor cells from human pluripotent stem cells (hPSCs). Cells are differentiated through four stages: 1) definitive endoderm, 2) primitive gut tube, 3) posterior foregut endoderm, and 4) pancreatic progenitor cells. The differentiated cells express key markers of pancreatic progenitor cells, including PDX-1, NKX6.1 and SOX9 and up-regulation of insulin and glucagon. The resulting pancreatic progenitor cells can be further differentiated to insulin-producing beta cells or other endocrine and exocrine pancreatic cell fates. STEMdiff™ Pancreatic Progenitor Kit has been optimised for differentiation of cells maintained in mTeSR™1 or mTeSR™ Plus.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').