MegaCult™ C Complete Kit with Cytokines
Catalog # STMC04971
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:MegaCult™-C Complete Kit with cytokines
Specifications
About this item
Complete kit including collagen, media, staining kit and chamber slides for human CFU-Mk assays.
MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining. MegaCult™-C medium is optimised for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples. It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').