MethoCult™ H4535 Enriched without EPO
STMC04545
:
- Förp.:24 Vial
- Beskrivning:MethoCult™ H4535 enriched without EPO
Pre-aliquoted methylcellulose-based medium with recombinant cytokines (without EPO) for human cells. 24×3 ml vials.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').