ContactUsLinkComponent
MethoCult™ H4435 Enriched Medium
MethoCult™ H4435 Enriched Medium
Katalog # STMC04435
Leverantör:  STEMCELL Technologies
CAS-nummer:  
undefined
MethoCult™ H4435 Enriched Medium
Katalog # STMC04435
Leverantör:  STEMCELL Technologies
CAS-nummer:  
Specifikationer
  • Förp.:
    100 mL
  • Beskrivning:
    MethoCult™ H4435 enriched
Specifikationer
Om denna produkt
Methylcellulose-based medium with recombinant cytokines for human cells. 1x100ml bottle.
MethoCult™ H4435 Enriched (MethoCult™ GF+ H4435) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. This medium supports optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage and megakaryocyte progenitor cells (CFU-GEMM). MethoCult™ H4435 Enriched is recommended for use with CD34+ cells and other purified cell populations, and for CFU assays performed as part of LTC-IC assays.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').