Specifikationer
- Förp.:100 mL
- Beskrivning:NeuroCult™ SM1 neuronal supplement
Specifikationer
Om denna produkt
Supplement (50X) for the serum-free culture of neurons.
Avoid culture failure with a Brewer’s B27-based supplement that provides a consistent neuronal culture experience for CNS-derived or human pluripotent stem cell (hPSC)-derived cells. NeuroCult™ SM1 (STEMCELL Modified-1) Neuronal Supplement is based on the published formulation and standardised to more reproducibly support survival and maturation of functional primary and human pluripotent stem cell (hPSC)-derived neurons. This serum-free supplement can be used with basal media and a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages. NeuroCult™ SM1 may also be used as a serum-replacement supplement for various customisable applications, such as neurotoxicity assays and calcium imaging. For convenience, NeuroCult™ SM1 is included as a component of multiple BrainPhys™ Neuronal Medium culture kits for primary and hPSC-derived neurons.