Orders LinkContactUsLinkComponent
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ endothelial differentiation kit, 1 KIT
Katalog # STMC08005
Marketsource
Leverantör:   Stemcell Technologies
STEMdiff™ Endothelial Differentiation Kit
Katalog # STMC08005
Leverantör:  Stemcell Technologies
Marketsource
CAS-nummer:  
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ Endothelial Differentiation Kit
STEMdiff™ endothelial differentiation kit, 1 KIT
CAS-nummer:  
Price
12 004,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Product Details & Documents
Serum- and xeno-free kit for differentiation and expansion of human pluripotent stem cell-derived endothelial cells.
STEMdiff™ endothelial differentiation kit includes attachment substrate, animal component-free (ACF) endothelial induction medium, and endothelial expansion medium. It is optimised for the differentiation of human pluripotent stem cells (hPSCs) maintained in mTeSR™1, mTeSR™ plus , or TeSR™-E8™ on Corning® Matrigel® to endothelial-like cells. The kit is designed to be used immediately after early mesoderm induction with STEMdiff™ mesoderm induction medium, available for purchase separately. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Förp.:
    1 KIT
  • Beskrivning:
    STEMdiff™ endothelial differentiation kit