Assistent® Brush for Beakers
Naturborsthår, 85 mm, Trä, 430 mm
Catalog # HECH41936010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Wood handle, natural bristle brush designed for use with beakers, or similar sized laboratory glassware.
- Wood handle
- Natural bristles
Natural bristles are ideal for cleaning beakers, glass or plastic cups, measuring cups and more.
Specifications
- Borstmaterial:Naturborsthår
- Borsthuvud Ø:85 mm
- Handle material:Trä
- Total längd:430 mm