Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:CD44 molecule (Indian blood group)
- Antigensymbol:CD44
- Klonalitet:Monoclonal
- Klon:HCM15-1
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG1 kappa
- Mått:Purified Antibody
- Form:Liquid
- Gen-ID:P16070
- Antigen synonymer:CD44
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Förvaringstemperatur:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Koncentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Purified human protein was used as the immunogen for the CD44 antibody.
- Rening:Protein G affinity chromatography
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Receptor for hyaluronic acid (HA). Mediates cell-cell and cell-matrix interactions through its affinity for HA, and possibly also through its affinity for other ligands such as osteopontin, collagens, and matrix metalloproteinases (MMPs). Adhesion with HA plays an important role in cell migration, tumor growth and progression. In cancer cells, may play an important role in invadopodia formation. Also involved in lymphocyte activation, recirculation and homing, and in hematopoiesis. Altered expression or dysfunction causes numerous pathogenic phenotypes. Great protein heterogeneity due to numerous alternative splicing and post-translational modification events. Receptor for LGALS9; the interaction enhances binding of SMAD3 to the FOXP3 promoter, leading to up-regulation of FOXP3 expression and increased induced regulatory T (iTreg) cell stability and suppressive function (By similarity). [UniProt].
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Titering of the CD44 antibody may be required for optimal performance.
1. FFPE testing requires sections to be boiled in pH6 10 mM citrate buffer for 10 - 20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCM15-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse