Anti-NOS3 Rabbit Polyclonal Antibody
Katalog # PRSI62-562
Leverantör: ProSci Inc.
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Nitric oxide synthase, endothelial
- Antigensymbol:NOS3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Western blot:Yes
- Epitop:48-75 aa
- Form:Liquid
- Gen-ID:4846
- Modifiering:None
- Molekylvikt:133 kDa
- Förvaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This NOS3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 48-75 amino acids from the N-terminal region of human NOS3.
- Rening:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Förp.:400 µl
Specifikationer
Om denna produkt
For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: NOS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 48-75
Host: Rabbit
Isotype: Ig
Reactivity: Human