Anti-TAF5 Rabbit Polyclonal Antibody
PRSI22-793
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:TATA-box binding protein associated factor 5
- Antigensymbol:TAF5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:6877
- Antigen synonymer:transcription initiation factor TFIID 100 kDa subunit|TAF(II)10TAF2D|TATA box binding protein (TBP)-associated factor 2D|TAF5 RNA polymerase II|TATA box binding protein (TBP)-associated factor|transcription initiation factor TFIID 100 kD subunit|100kDa|TAFII-10TAFII10transcription initiation factor TFIID subunit 5|RNA polymerase II|100kD|D
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 656-800 of human TAF5 (NP_008882.2)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: TAF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat