Anti-SAT2 Rabbit Polyclonal Antibody
PRSI22-787
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Spermidine/spermine N1-acetyltransferase family member 2
- Antigensymbol:SAT2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:112483
- Antigen synonymer:diamine N-acetyltransferase 2|SSAT2|thialysine N-epsilon-acetyltransferase|spermidine/spermine N(1)-acetyltransferase 2|diamine acetyltransferase 2|polyamine N-acetyltransferase 2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human SAT2 (NP_597998.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: SAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse