Anti-PSMD5 Rabbit Polyclonal Antibody
Catalog # PRSI22-584
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:26S proteasome non-ATPase regulatory subunit 5
- Antigensymbol:PSMD5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:5711
- Antigen synonymer:26S proteasome non-ATPase regulatory subunit 5|26S proteasome subunit S5B|26S protease subunit S5 basic|KIAA0072|PSMD5
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 255-504 of human PSMD5 (NP_005038.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: PSMD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat