Anti-POLR2H Rabbit Polyclonal Antibody
Catalog # PRSI22-578
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Polymerase (RNA) II (DNA directed) polypeptide H
- Antigensymbol:POLR2H
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Immunutfällning:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:5437
- Antigen synonymer:RPB8|RPB17|RPABC3|hsRPB8|POLR2H
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human POLR2H (NP_006223.2)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: POLR2H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat