Anti-MYO1C Rabbit Polyclonal Antibody
Katalog # PRSI22-567
Leverantör: ProSci Inc.
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Unconventional myosin-Ic
- Antigensymbol:MYO1C
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:4641
- Antigen synonymer:Myosin I beta|MYO1C|Unconventional myosin-Ic|MMI-beta|MMIb
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 769-1028 of human MYO1C (NP_203693.3)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: MYO1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat