Anti-AFF1 Rabbit Polyclonal Antibody
PRSI22-565
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:AF4/FMR2 family, member 1
- Antigensymbol:AFF1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gen-ID:4299
- Antigen synonymer:AF4/FMR2 family member 1|MLLT2|AF4|Proto-oncogene AF4|PBM1|AFF1|Unknown|Protein AF-4|member 1|FEL|Protein FEL|AF4/FMR2 family|AF-4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1124-1218 of human AFF1 (NP_001160165.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: AFF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat