Anti-KLRB1 Rabbit Polyclonal Antibody
PRSI22-562
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Killer cell lectin-like receptor subfamily B, member 1
- Antigensymbol:KLRB1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:3820
- Antigen synonymer:NKRP1A|Killer cell lectin-like receptor subfamily B member 1|hNKR-P1A|NKR-P1A|NKR-P1|NKR|C-type lectin domain family 5 member B|CLEC5B|CD161
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 73-140 of human KLRB1 (NP_002249.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: KLRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat