Anti-HIP1 Rabbit Polyclonal Antibody
Catalog # PRSI22-556
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Huntingtin interacting protein 1
- Antigensymbol:HIP1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:3092
- Antigen synonymer:HIP-I|HIP-1|ILWEQ|HIP1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 410-690 of human HIP1 (NP_005329.3)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: HIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse