Anti-ASPH Rabbit Polyclonal Antibody
Catalog # PRSI22-521
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Aspartate beta-hydroxylase
- Antigensymbol:ASPH
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:444
- Antigen synonymer:CASQ2BP1|BAH|AAH|HAAH|JCTN|ASPH|junctin
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 81-270 of human ASPH (NP_001158227.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: ASPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human