Anti-PPP1R8 Rabbit Polyclonal Antibody
Catalog # PRSI22-418
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Protein phosphatase 1, regulatory subunit 8
- Antigensymbol:PPP1R8
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:5511
- Antigen synonymer:PRO2047|NIPP1|NIPP-1|ARD1|ARD-1|PPP1R8
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-209 of human PPP1R8 (NP_612568.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: PPP1R8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat