Anti-PPARA Rabbit Polyclonal Antibody
PRSI22-414
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Peroxisome proliferator-activated receptor alpha
- Antigensymbol:PPARA
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:5465
- Antigen synonymer:PPARA|PPAR|PPARalpha|NR1C1|hPPAR
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human PPAR alpha (NP_001001928.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: PPARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat