Anti-OGN Rabbit Polyclonal Antibody
PRSI22-401
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Mimecan
- Antigensymbol:OGN
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:4969
- Antigen synonymer:Osteoinductive factor|OGN|Mimecan|SLRR3A|Osteoglycin|OIF
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-298 of human OGN (NP_148935.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: OGN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat