Anti-KCNH1 Rabbit Polyclonal Antibody
PRSI22-369
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Potassium voltage-gated channel subfamily H member 1
- Antigensymbol:KCNH1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:3756
- Antigen synonymer:voltage-gated potassium channel subunit Kv10.1|EAG channel 1|Drosophila|potassium voltage-gated channel|ZLS1|hEAG1|EAG|ether-a-go-go potassium channel 1|EAG1|potassium channel|voltage gated eag related subfamily H|ether-a-go-go|h-eag|potassium voltage-gated channel subfamily H member 1|member 1|Kv10.1|homolog of|TMBTS|subfamily H (eag-related)
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 813-962 of human KCNH1 (NP_002229.1)
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Type: Primary
Antigen: KCNH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat