Anti-TPD52L1 Rabbit Polyclonal Antibody
PRSI60-103
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:Tumor protein D53
- Antigensymbol:TPD52L1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:43 - 70 aa
- Form:Liquid
- Gen-ID:7164
- Modifiering:None
- Molekylvikt:22 kDa
- Förvaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Koncentration:0,42 mg/ml
- Shipping temperature:4°C
- Immunogen:This TPD52L1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 43 - 70 amino acids from the N-terminal region of human TPD52L1.
- Rening:This antibody is purified through a protein A column, followed by peptide affinity purification
- Förp.:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: TPD52L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 43-70
Host: Rabbit
Isotype: Ig
Reactivity: Mouse