Anti-NOSIP Rabbit Polyclonal Antibody
Catalog # PRSI59-298
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Nitric oxide synthase-interacting protein
- Antigensymbol:NOSIP
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:67 - 95 aa
- Form:Liquid
- Gen-ID:51070
- Modifiering:None
- Molekylvikt:33 kDa
- Förvaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Koncentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This NOSIP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 67 - 95 amino acids from the N-terminal region of human NOSIP.
- Rening:This antibody is purified through a protein A column, followed by peptide affinity purification
- Förp.:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: NOSIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 67-95
Host: Rabbit
Isotype: Ig
Reactivity: Human