Anti-NOS2 Rabbit Polyclonal Antibody
Katalog # PRSI57-527
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Nitric oxide synthase, inducible
- Antigensymbol:NOS2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:108-138 aa
- Form:Liquid
- Gen-ID:4843
- Modifiering:None
- Molekylvikt:131 kDa
- Förvaringstemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Koncentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NOS2A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 108-138 amino acids from the N-terminal region of human NOS2A
- Rening:This antibody is purified through a protein A column, followed by peptide affinity purification
- Förp.:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: NOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 108-138
Host: Rabbit
Isotype: Ig
Reactivity: Human