Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:CD3e molecule, epsilon (CD3-TCR complex)
- Antigensymbol:CD3E
- Klonalitet:Monoclonal
- Klon:UCHT1
- Konjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flödescytometri:Yes
- Värd:Mouse
- Isotyp:IgG1 kappa
- Korsad adsorption:No
- Form:liquid
- Gen-ID:P07766
- Antigen synonymer:CD3E
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Förvaringstemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Rening:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Förp.:0,025 mg
Specifications
About this item
The UCHT1 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3 epsilon. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human