Anti-CD3E Mouse Monoclonal Antibody [clone: OKT3] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Katalog # PRSI76-252
Leverantör: ProSci Inc.
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:CD3e molecule, epsilon (CD3-TCR complex)
- Antigensymbol:CD3E
- Klonalitet:Monoclonal
- Klon:OKT3
- Konjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flödescytometri:Yes
- Värd:Mouse
- Isotyp:IgG2a kappa
- Reaktivitet:Human
- Korsad adsorption:No
- Form:liquid
- Gen-ID:P07766
- Antigen synonymer:CD3E
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Förvaringstemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Rening:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Förp.:25 Tests
Specifikationer
Om denna produkt
The OKT3 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The OKT3 antibody is an immunosuppressive, which has proven to be an effective therapeutic agent in liver, heart, and renal allograft rejection.
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human