Anti-Il7r Rat Monoclonal Antibody [clone: A7R34] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # PRSI76-918
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:interleukin 7 receptor
- Antigensymbol:Il7r
- Klonalitet:Monoclonal
- Klon:A7R34
- Konjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flödescytometri:Yes
- Värd:Rat
- Isotyp:IgG2a kappa
- Reaktivitet:Mouse
- Korsad adsorption:No
- Form:liquid
- Gen-ID:P16872
- Antigen synonymer:Il7r
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Förvaringstemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Rening:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Förp.:0,025 mg
Specifications
About this item
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the alpha subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.
Type: Primary
Antigen: Il7r
Clonality: Monoclonal
Clone: A7R34
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse