Anti-PTPRC Mouse Monoclonal Antibody [clone: HI100] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
PRSI76-460
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:protein tyrosine phosphatase, receptor type, C
- Antigensymbol:PTPRC
- Klonalitet:Monoclonal
- Klon:HI100
- Konjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flödescytometri:Yes
- Värd:Mouse
- Isotyp:IgG2b kappa
- Korsad adsorption:No
- Form:liquid
- Gen-ID:P08575
- Antigen synonymer:PTPRC
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Förvaringstemperatur:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Rening:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Förp.:25 Tests
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI100
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human