Anti-GLB1 Recombinant Antibody [Clone: 13R4]
Catalog # PRSI24-140
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:galactosidase beta 1
- Antigensymbol:GLB1
- Klonalitet:Recombinant
- Klon:13R4
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Isotyp:Fab fragment lambda
- Reaktivitet:E. coli
- Western blot:Yes
- Korsad adsorption:No
- Form:Liquid
- Gen-ID:2720
- Antigen synonymer:MPS4B| Acid beta galactosidase|EBP|β-galactosidase|β-gal|GLB1|beta-gal|ELNR1|Galactosidase beta 1|Beta galactosidase 1
- Modifiering:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Förvaringstemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Beta-galactosidase
- Tested applications:Negative control
- Förpackningstyp:Vial
- Förp.:0,2 mg
Specifications
About this item
Type: Primary
Antigen: galactosidase beta 1
Clonality: Recombinant
Clone: 13R4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Fab fragment lambda
Reactivity: E. coli