Anti-DM1 Mouse Monoclonal Antibody [clone: 15L2]
Katalog # GENSA02223-40
Leverantör: Genscript
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:DM1
- Klonalitet:Monoclonal
- Klon:15L2
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- Isotyp:IgG1 kappa
- Korsad adsorption:No
- Form:Liquid
- Storage buffer:Supplied in PBS, pH 7,4, containing 0,02% ProClin300
- Förvaringstemperatur:The product remains stable up to 1 year at −20 °C from date of receipt. Avoid repeated freezing and thawing cycles.
- Koncentration:0,5 mg/ml
- Immunogen:DM1-KLH
- Förpackningstyp:Vial
- Förp.:40 µG
Specifikationer
Om denna produkt
The DM1, also known as mertansine, is a microtubule polymerisation inhibitor. Trastuzumab emtansine (T-DM1), composed of trastuzumab conjugated to DM1, is a type of targeted cancer drug for breast cancer treatment. T-DM1 is targeted toward HER2 (ERBB2) positive breast cancer.
- High specificity
- High affinity
- High stability