2-Chlorostyrene
Catalog # MOLE64920222-5G
Supplier: Molekula
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:5 g
- Seq:3173
Specifications
About this item
:
:138.6 g/mol
Boiling Pt:188,7 °C (1013 hPa)
Melting Pt:–63,2 °C
Density:1,10001 g/cm³ (20 °C)
EINECS:218-026-8
CAS Number:2039-87-4
Formula:C₈H₇Cl
MDL Number:MFCD00000567