


HPF ~5mM ≥98%
Catalog # ENZOALX620074M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 mg
- Seq:2070
Specifications
Specification Test Results
- Concentration:~5 mM
- Formulation:Dissolved in 0.47 ml dimethyl formamide
- Purity:≥98% (HPLC)
Specification Test Results
About this item
:
:424.4 g/mol
Storage Temperature:Kylskåp
EINECS:200-679-5
UN:2265
ADR:3,III
:GHS02
:GHS07
:GHS08
CAS Number:359010-69-8
Formula:C₂₆H₁₆O₆