ContactUsLinkComponent
Vial Tray
Vial Tray
Katalog # MOFI234008
Vial Tray
Katalog # MOFI234008
Specifikationer
  • Beskrivning:
    Tray with plastic gate, white
  • B×D×H:
    375×375×70 mm
  • Förp.:
    1
Specifikationer
Om denna produkt
Stackable vial tray.
  • Tapered front lip for easy loading
  • Smooth, non-porous surface is easy to clean
Made of polypropylene for normal use in temperatures less than 82 °C.