Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standarder
Turbiditetsstandarder
McFarland Barium Sulphate Turbidity Standard
McFarland Barium Sulphate Turbidity Standard
McFarland Barium Sulphate Turbidity Standard
Catalog # LIOF80402
Supplier:
LIOFILCHEM
CAS Number:
Expand image
McFarland Barium Sulphate Turbidity Standard
Catalog # LIOF80402
Supplier:
LIOFILCHEM
CAS Number:
Specifications
Förp.:
1
Specifications
About this item
Product Details
Product Details