KIMBLE® Black Phenolic, Screw Thread Caps
Katalog # WHEA73809-15425
Leverantör: DWK Life Sciences
Specifikationer
- Lock storlek:15-425
- Färg:Svart
- Beskrivning:Black phenolic, screw thread caps, with LDPE cone liner, Cap style: solid top
- Material:Phenolic resin
- Förp.:144
Specifikationer
Om denna produkt
Black phenolic cap has a securely mounted LDPE cone-shaped liner which offers a two part seal. The closure forms a seal around the rim and the polyseal cone forms a seal against the inner diameter of the vial opening.
- Designed for superior torque retention
- Stress crack resistant
- Excellent for sample storage and re-sealing
- Identified in ASTM Specification E982, Class A requirements
- Continuous knurling