WHEATON® Screw Closures, Polypropylene
Catalog # WHEAW015324
Supplier: DWK Life Sciences
Specifications
- Gänga:13-425
- Färg:Vit
- Beskrivning:Screw closures, solid top, PTFE
- Material:PP
- Förp.:1
Specifications
About this item
Use these Polypropylene caps with glass/plastic bottles or vials.
- Polyvinyl liners can be used with mild acids, alkalis, solvents and alcohols
- Foamed polyethylene (PE) liner provides chemical resistance for acids, alkalis, solvents and alcohols
PTFE faced foamed PE liner ideal for use with high purity chemicals, strong acids and solvents.