Orders LinkContactUsLinkComponent
WHEATON® Snap On Stopper
WHEATON® Snap On Stopper
Snap on stopper, grey chlorobutyl, for mouth ext.Ø 20 mm, 45 durometer
Katalog # WHEAW015258
Leverantör:   DWK Life Sciences
WHEATON® Snap On Stopper
Katalog # WHEAW015258
Leverantör:  DWK Life Sciences
WHEATON® Snap On Stopper
Snap on stopper, grey chlorobutyl, for mouth ext.Ø 20 mm, 45 durometer
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 1.000 Items
Product Details & Documents
Snap-on stopper provides maximum sealing for vials by locking into the blowback ridge.
Provides a tight seal and keeps the stopper from backing out.
Specifikationer
  • Beskrivning:
    Snap on stopper, grey chlorobutyl, for mouth ext.Ø 20 mm, 45 durometer