Electronic Overhead Stirrers, Hei-TORQUE Platinum Package
Katalog # HEID501-64029-04
Leverantör: Heidolph Instruments GmbH & Co.KG
Specifikationer
- Beskrivning:Hei-TORQUE Platinum package, CH plug
- Förp.:1 SET
Specifikationer
Om denna produkt
Hei-TORQUE Platinum package with the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory.
- Hei-TORQUE Ultimate 400
- Telescope stand
- Clamp
Packaging: Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.