Boss Head
Catalog # 431-0043
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Beskrivning:Cross sleeve (2 clamps are required to attach the SilentCrusher M and the vessel holding bracket)
- Förp.:1
Specifications
About this item
Boss head made of anodised aluminum, used to fix retort clamps to stand rods for perfect vertical alignment.
Suitable for stand rods with up to 13,1 mm maximum diameter.