Specifications
- Handskstorlek:XL
- Ej handspecifika/handspecifika:Handspecifika
- Engångs/Återanvändbar:Återanvändbar
- Färg:Svart
- EN standard:EN ISO 388:2016 (4X31A)
- Kragmodell:Stickad mudd
- Fodermaterial:Nylon
- Mätare:15
- Yta:Nitrile micro-foam black coating
- Förp.:10
Specifications
About this item
All in one glove with enhanced cut resistance, comfort, ease of selection, cost effectiveness. Due to its high-performance fiber, the Honeywell CoreShield™ range of 22 styles offers complete cut protection from level A/A1 to F/A9, in a choice of four various gauges (from 10 gg to 18 gg) and three different coatings.
- Easy to wear
- Thumb reinforcement
- Touchscreen capability for resistive screen machines
- Silicone-free
- Hand and wrist protection
CoreShield™ High Performance micro-foam nitrile coating provides 360° breathability, excellent grip performance, water and oil proof coating, abrasion resistance, reduced hand fatigue and excellent dexterity.
CoreShield™ Engineered Yarn: Skin friendly and comfortable material to shield the cut resistant material. Softer skin-friendly yarn in contact with the skin.
Industries: Manufacturing, automotive, metal stamping, glass handling, construction, logistics.