Stoppers, Glass, Conical Ground Joint
Katalog # 217-0767
Leverantör: DWK Life Sciences
Specifikationer
- NS:19/26
- Typ:Clear glass
- Förp.:10
Specifikationer
Om denna produkt
DURAN® glass stopper with conical ground joint, for reagent bottles and oxygen bottles according to Winkler.
- Made of borosilicate 3.3 glass